missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55017-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to TSC22D4.
ZNF289 (zinc finger protein 289), also known as ARFGAP2 (ADP-ribosylation factor GTPase activating protein 2), IRZ, Zfp289 or Nbla10535, functions as a GTPase-activating protein (GAP) for ARF family proteins. Localizing to the cytoplasmic side of the Golgi apparatus, ZNF289 contains one ARF-GAP domain and is found associated with COP-I-coated vesicles.Spécification
TSC22D4 | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3B8N7, Q9EQN3, Q9Y3Q8 | |
THG 1, THG1, TILZ2, Tsc 22 like protein THG 1, TSC22 domain family protein 4, TSC22 domain family, member 4, TSC22D4 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
684980, 78829, 81628 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
TSC22D4 | |
Liquid | |
Tsc22d4 | |
Peptide | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |