Learn More
TSH-Receptor, B-Chain Mouse, Clone: TSHRB/1405, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 7253-MSM5-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
TSH-Receptor, B-Chain | |
Cell Surface and Cytoplasmic | |
TSHRB/1405 | |
Unconjugated | |
XP_005268094 | |
Mouse | |
42kDa | |
Protein A/G Purified | |
RUO | |
7253 | |
IgG2a |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
TSHR | |
Recombinant human TSHR, B-Chain protein | |
293T Cells. Thyroid gland | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |