missing translation for 'onlineSavingsMsg'
Learn More

TSPAN15 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10460-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16896853

  • 349.00€ / 150µL
Expédition estimée: 29-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
Spécification

Spécification

TSPAN15
Polyclonal
Unconjugated
TSPAN15
2700063A19Rik, NET 7, NET7, Tetraspan NET 7, tetraspanin 15, TM4SF15, Tspan 15, TSPAN15
Rabbit
Antigen Affinity Chromatography
RUO
23555
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95858
TSPAN15
TSPAN15 Fusion Protein Ag0710
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions