missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC39B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23432-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
There are 2 named isoforms produced by alternative splicing.Spécification
TTC39B | |
Polyclonal | |
Unconjugated | |
TTC39B | |
C9orf52, TPR repeat protein 39B, TTC39B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158219, 69863 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5VTQ0, Q8BYY4 | |
TTC39B | |
TTC39B Fusion Protein Ag20081 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |