missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13618-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.Spécification
TTL | |
Polyclonal | |
Unconjugated | |
TTL | |
TTL, Tubulin tyrosine ligase, tubulin tyrosine ligase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
150465, 171572, 69737 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P38585, Q8NG68, Q9QXJ0 | |
Ttl | |
TTL Fusion Protein Ag4526 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |