missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16016-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Mediates thiol-dependent retention in the early secretory pathway, forming mixed disulfides with substrate proteins through its conserved CRFS motif. Inhibits the calcium channel activity of ITPR1. May have a role in the control of oxidative protein folding in the endoplasmic reticulum. Required to retain ERO1L and ERO1LB in the endoplasmic reticulum.Spécification
TXNDC4 | |
Polyclonal | |
Unconjugated | |
ERP44 | |
ER protein 44, ERP44, KIAA0573, PDIA10, TXNDC4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23071 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BS26 | |
ERP44 | |
TXNDC4 Fusion Protein Ag8818 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |