Learn More
TYK2 Mouse anti-Human, Unconjugated, Clone: 1E1A10, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67411-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830026
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
Description
The TYK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TYK2. This antibody recognizes human antigen. The TYK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
TYK2
Monoclonal
800 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1E1A10
Unconjugated
150 μL
7297
IgG2b