missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U2AF35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10334-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene belongs to the splicing factor SR family of genes. U2 auxiliary factor, comprising a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the small subunit which plays a critical role in both constitutive and enhancer-dependent RNA splicing by directly mediating interactions between the large subunit and proteins bound to the enhancers. Alternatively spliced transcript variants encoding different isoforms have been identified.Spécification
U2AF35 | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q01081, Q9D883 | |
FP793, RN, RNU2AF1, U2AF1, U2AF35, U2AFBP | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
108121, 687575, 7307 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
U2AF1 | |
Liquid | |
U2AF1 | |
U2AF35 Fusion Protein Ag0399 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |