missing translation for 'onlineSavingsMsg'
Learn More

UBC9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10224-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806473

  • 329.85€ / 150µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

In heart and muscle extracts the UBE2I antibodies recognizes a 38 kDa protein band, but this band is not visible in extracts of other rat tissues.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
Spécification

Spécification

UBC9
0.42 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P63279, P63280, P63281
C358B7.1, E2,UBE2I, P18, SUMO conjugating enzyme UBC9, SUMO protein ligase, UBC9, UBCE9, UBE2I, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin protein ligase I
Rabbit
IgG
150 μL
-20°C
Primary
22196, 25573, 7329
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
UBE2I
Liquid
UBE2I
UBC9 Fusion Protein Ag0252
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents