missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21951-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Urocortin (UCN), a 40 amino acids neuropeptide widely distributed in the central nervous system (CNS), belongs to a family of structurally related peptides including corticotrophin releasing factor (CRF), sauvagine and urotensin I. Urocortin is derived from a larger precursor protein (pro-urocortin, also named corticotensin). Urocortin has biological actions similar to CRF such as stress-related endocrine, autonomic and behavioral responses in mammalian species. It is a potent stimulator of adrenocorticotropic hormone (ACTH) secretion by corticotroph cells from the anterior pituitary in vitro and in vivo.Spécification
UCN | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55089 | |
UCN, UI, UROC, urocortin | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
7349 |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
UCN | |
Liquid | |
UCN | |
UCN Fusion Protein Ag15995 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |