Learn More
Abeba™ UNI6 36762 Shoes
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
Marque:  Abeba™ 36762/36
Code nomenclature Nacres: HA.03
Informations supplémentaires : Poids : 0.98000kg
 Afficher plus de versions de ce produit
Code produit. 15660568
  • 90.60€ / 1 paire
Expédition estimée
10-10-2019
Ajouter au panier
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Functional leather with honeycomb pattern
  • Blue
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Scratch-proof
  • Water-repellent
Spécification
Spécification
CE EN ISO 20347:2012 O1 FO SRC
Leather
Slip resistant
36762
Blue
36
Polyurethane