Learn More
Simport™ Scientific UNISETTE™ Biopsy Processing/Embedding Cassette
Secure specimens during processing with Simport Scientific™ UNISETTE™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
Marque:  Simport™ Scientific M506-6
Code nomenclature Nacres: NB.17
Informations supplémentaires : Poids : 1.00000kg
Code produit. 15132964
Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 03 88 67 14 14.
Description
Description
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes
Certifications
CE Marked
Spécification
Spécification
Biopsy Processing/Embedding Cassette
35°
Blue
Acetal
Vidéos
For in vitro diagnostic (IVD) use only.