Learn More
Simport™ Scientific UNISETTE™ Biopsy Processing/Embedding Cassette
Secure specimens during processing with Simport Scientific™ UNISETTE™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
Marque:  Simport™ Scientific M506-6
Code nomenclature Nacres: NB.17
Informations supplémentaires : Poids : 1.00000kg
 Afficher plus de versions de ce produit
Code produit. 15132964
  • 199.00€ / 1 pièce
Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier
Avertissements:
For IVD use
Description
Description
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes
Certifications
CE
Spécification
Spécification
Biopsy Processing/Embedding Cassette
35°
Blue
Acetal
Vidéos