missing translation for 'onlineSavingsMsg'
Learn More

Acide boronique d’uracil-5-, 95 %, Thermo Scientific Chemicals

Catalog Number 11352209
Click to view available options
Quantité:
250 mg
1 g
70523-22-7
C4H5BN2O4
155.90
MFCD01318983
PVEJOCQTIVCDNO-UHFFFAOYSA-N
uracil-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl boronic acid, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 2,4-dihydroxypyrimidine-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-ylboronic acid, 2,4-dihydroxypyrimidin-5-ylboronic acid, 2,4-dioxo-1h-pyrimidin-5-yl boronic acid, 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid, t6mvmvj ebqq wln, 5-dihydroxyboryluracil
256205
(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
OB(O)C1=CNC(=O)NC1=O
70523-22-7
C4H5BN2O4
155.90
MFCD01318983
PVEJOCQTIVCDNO-UHFFFAOYSA-N
uracil-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl boronic acid, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 2,4-dihydroxypyrimidine-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-ylboronic acid, 2,4-dihydroxypyrimidin-5-ylboronic acid, 2,4-dioxo-1h-pyrimidin-5-yl boronic acid, 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid, t6mvmvj ebqq wln, 5-dihydroxyboryluracil
256205
(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)boronic acid
OB(O)C1=CNC(=O)NC1=O

It finds its application in the probing the interactions between boronic acids and cis-diol-containing biomolecules by affinity capillary electrophoresis. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trouve son application dans l’analyse des interactions entre les acides boroniques et les biomolécules contenant du cis-diol par électrophorèse capillaire d’affinité. Il sert aussi d’intermédiaire pharmaceutique actif.

Solubilité
Légèrement soluble dans l’eau.

Remarques
Conserver à l’abri des oxydants. Gardez le récipient bien fermé et placez-le dans un endroit frais, sec et bien ventilé.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Uracil-5-boronic acid
Point de fusion >300°C
Quantité 1 g
Informations sur la solubilité Slightly soluble in water.
Poids de la formule 155.9
Pourcentage de pureté 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'