missing translation for 'onlineSavingsMsg'
Learn More
Learn More
URM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15285-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Following the discovery of protein modification by the small, highly conserved ubiquitin polypeptide, a number of distinct ubiquitin-like proteins (Ubls) have been found to function as protein modifiers as well. These Ubls, which include SUMO, ISG15, Nedd8, and Atg8, function as critical regulators of many cellular processes, including transcription, DNA repair, signal transduction, autophagy, and cell-cycle control. A growing body of data also implicates the dysregulation of Ubl-substrate modification and mutations in the Ubl-conjugation machinery in the etiology and progression of a number of human diseases. URM1 is a ubiquitin-like modifier protein.Spécification
URM1 | |
Polyclonal | |
Unconjugated | |
URM1 | |
C9orf74, URM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311840, 68205, 81605 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BTM9, Q9D2P4 | |
Urm1 | |
URM1 Fusion Protein Ag7448 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |