missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UROD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15547-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes the fifth enzyme of the heme biosynthetic pathway. This enzyme is responsible for catalyzing the conversion of uroporphyrinogen to coproporphyrinogen through the removal of four carboxymethyl side chains. Mutations and deficiency in this enzyme are known to cause familial porphyria cutanea tarda and hepatoerythropoetic porphyria.Spécification
UROD | |
Polyclonal | |
Unconjugated | |
UROD | |
PCT, UPD, URO D, UROD, uroporphyrinogen decarboxylase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22275, 29421, 7389 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06132, P32362, P70697 | |
UROD | |
UROD Fusion Protein Ag7905 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |