missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22327-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the basic helix-loop-helix leucine zipper family, and can function as a cellular transcription factor. The encoded protein can activate transcription through pyrimidine-rich initiator (Inr) elements and E-box motifs. This gene has been linked to familial combined hyperlipidemia (FCHL). Two transcript variants encoding distinct isoforms have been identified for this gene.Spécification
USF1 | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22415 | |
bHLHb11, FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, Upstream stimulatory factor 1, USF, USF1 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
7391 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
USF1 | |
Liquid | |
USF1 | |
USF1 Fusion Protein Ag17789 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |