missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP13 Mouse anti-Human, Clone: 6E8C10, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66176-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
USP13, also known as Ubiquitin carboxyl-terminal hydrolase 13, is a deubiquitinating enzyme involved in ubiquitin-dependent protein catabolism. A member of the peptidase C19 family, USP13 is a highly conserved protein and contains two UBA domains and a UBP-type zinc finger. Expression of USP13 is higher in ovary and testis.Spécification
USP13 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q92995 | |
USP13 | |
USP13 Fusion Protein Ag10506 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
6E8C10 | |
Unconjugated | |
USP13 | |
Deubiquitinating enzyme 13, Isopeptidase T 3, IsoT 3, ISOT3, Ubiquitin thiolesterase 13, USP13 | |
Mouse | |
Protein G | |
RUO | |
8975 | |
-20°C | |
Liquid |