missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP44 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15521-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP44 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes.[supplied by OMIM, Mar 2008].Spécification
USP44 | |
Polyclonal | |
Unconjugated | |
USP44 | |
Deubiquitinating enzyme 44, Ubiquitin thiolesterase 44, USP44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
327799, 84101 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C2S0, Q9H0E7 | |
Usp44 | |
USP44 Fusion Protein Ag7888 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |