Learn More
Binder™ Chambres de séchage sous vide, série VD
Ensures optimal heat transfer with patented Binder™ expansion shelf technology. Binder™ Vacuum Drying Chamber, Series VD uses homogenous temperature distribution and includes shelves that are easy to position.
7712.00€ - 10675.00€
Produits 2
Code produit Marque Dimensions (L x l x H) extérieures Prix Quantité et disponibilité  
Code produit Marque Dimensions (L x l x H) extérieures Prix Quantité et disponibilité  
15642186 Binder™
9030-0030
7712.00€
1 pièce
Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier
15652186 Binder™
9030-0031
10675.00€
1 pièce
Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier
Description
Description
  • Fast and gentle drying
  • Direct heat transfer through large thermal conducting plates
  • Safe work with one-of-a-kind safety concept
  • Coordinated, modular system with vacuum pump and vacuum module
  • Temperature range: room temperature plus 15 °C to 200 °C
  • APT.line™ preheating chamber technology
  • Controller with time-segment and real-time programming
  • 2 aluminum expansion racks
  • Inert gas connection
  • Shatterproof, spring-mounted safety glass panel
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Computer interface: RS 422
  • 2 relay outputs, 24 V DC (max. 0.4 A)