Learn More
VPS35 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10236-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The VPS35 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPS35. This antibody recognizes human, mouse, rat antigen. The VPS35 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Spécification
VPS35 | |
Polyclonal | |
Unconjugated | |
20 μL | |
55737 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP) | |
260 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |