missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15079-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Plays a role in vesicle-mediated protein trafficking of the endocytic membrane transport pathway. Believed to act as a component of the putative CORVET endosomal tethering complexes which is proposed to be involved in the Rab5-to-Rab7 endosome conversion probably implicating MON1A/B, and via binding SNAREs and SNARE complexes to mediate tethering and docking events during SNARE-mediated membrane fusion. The CORVET complex is proposed to function as a Rab5 effector to mediate early endosome fusion probably in specific endosome subpopulations. Functions predomitly in APPL1-containing endosomes.Spécification
VPS8 | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q0P5W1, Q8N3P4 | |
KIAA0804, VPS8 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
209018, 23355, 287990 |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
VPS8 | |
Liquid | |
Vps8 | |
VPS8 Fusion Protein Ag7151 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |