missing translation for 'onlineSavingsMsg'
Learn More

VTA1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15786-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805124

  • 329.85€ / 150µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

C6ORF55 encodes a protein involved in trafficking of the multivesicular body, an endosomal compartment involved in sorting membrane proteins for degradation in lysosomes.
Spécification

Spécification

VTA1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NP79
C6orf55, DRG 1, DRG1, HSPC228, LIP5, LYST interacting protein 5, My012, SBP1, SKD1 binding protein 1, VTA1
Rabbit
IgG
150 μL
-20°C
Primary
51534
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
VTA1
Liquid
VTA1
VTA1 Fusion Protein Ag8485
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents