missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vwf Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27186-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.Spécification
vwf | |
Polyclonal | |
Unconjugated | |
VWF | |
F8VWF, von Willebrand antigen II, von Willebrand factor, VWD, vwf | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7450 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04275 | |
VWF | |
vwf Fusion Protein Ag25578 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |