Learn More
WDR12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA557635
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: MAQLQTRFYT DNKKYAVDDV PFSIPAASEI ADLSNIINKL LKDKNEFHKH VEFDFLIKGQ FLRMPLDKHM EMENISSEEV VEI Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
WDR12 Polyclonal antibody specifically detects WDR12 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Spécification
WDR12 | |
Polyclonal | |
Unconjugated | |
WDR12 | |
4933402C23Rik; fb24f09; HAMAP-Rule:MF_03029}; MNCb-5414; nuclear protein Ytm1; ribosome biogenesis protein wdr12; ribosome biogenesis protein WDR12 {ECO:0000255; WD repeat domain 12; WD repeat domain 12 protein; WD repeat-containing protein 12; WD repeat-containing protein 12 {ECO:0000255; WDR12; wdr12 protein; wu:fb24f09; YTM1; Ytm1p; zgc:55609 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55759, 57750 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZL7, Q9JJA4 | |
WDR12 | |
Recombinant protein corresponding to Human WDR12 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only