missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27628-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Studies of the related mouse protein suggest that the encoded protein may play a role in the process of ossification.Spécification
WDR8 | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P2S5 | |
WD repeat containing protein 8, WD repeat domain 8, WD repeat-containing protein WRAP73, WDR8, WRAP73 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
49856 |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
WRAP73 | |
Liquid | |
WRAP73 | |
WDR8 Fusion Protein Ag26267 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |