missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WFDC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24917-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains four WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Alternatively spliced transcript variants have been observed but their full-length nature has not been determined.Spécification
WFDC3 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IUB2 | |
WAP14, WFDC3 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
140686 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
WFDC3 | |
Liquid | |
WFDC3 | |
WFDC3 Fusion Protein Ag20207 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |