Learn More
WIPI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28820-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16836245
  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
Description
The WIPI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WIPI2. This antibody recognizes human, mouse antigen. The WIPI2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
WIPI2
Polyclonal
Unconjugated
150 μL
26100
IgG
Immunofluorescence, Immunohistochemistry, Western Blot
300 μg/mL
Rabbit
Primary
Mouse, Human