missing translation for 'onlineSavingsMsg'
Learn More

XDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 55156-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16806635

  • 329.85€ / 150µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to XDH.

XDH belongs to the group of molybdenum-containing hydroxylases involved in the oxidative metabolism of purines. The enzyme is a homodimer. This protein can be converted to xanthine oxidase by reversible sulfhydryl oxidation or by irreversible proteolytic modification. Defects in xanthine dehydrogenase cause xanthinuria, may contribute to adult respiratory stress syndrome, and may potentiate influenza infection through an oxygen metabolite-dependent mechanism.
Spécification

Spécification

XDH
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22985, P47989, Q00519
xanthine dehydrogenase, Xanthine Oxidase, Xanthine oxidoreductase, XD, XDH, XDHA, XO, XOR
Rabbit
IgG
150 μL
-20°C
Primary
22436, 497811, 7498
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
XDH
Liquid
Xdh
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents