Learn More
Waters Corporation XTerra MS C8 Column
XTerra MS C8 columns are general purpose, hybrid-based, reversed-phase C8 columns that are designed to be compatible with mass spectrometry applications.
Marque: Waters Corporation 186000415
Informations supplémentaires : Poids : 0.50000kg
Description
- These first generation columns provide superior pH stability compared to a silica-based column
Spécification
3 mm | |
6000 psi (415 Bar) | |
3.5 μm | |
1/Ea. | |
Yes |
50 mm | |
Spherical | |
125 Å | |
L7 | |
Column |