missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22944-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a nuclear-localized protein that may be induced by p53 and regulates the cell cycle by inhibiting G1 to S phase progression. The encoded protein may interact with other cell cycle regulators.Spécification
ZBTB10 | |
Polyclonal | |
Unconjugated | |
ZBTB10 | |
RINZF, RINZFC, ZBTB10, Zinc finger protein RIN ZF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229055, 65986 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96DT7 | |
ZBTB10 | |
ZBTB10 Fusion Protein Ag19094 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |