missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB8A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24544-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB8, also known as BOZ-F1, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB8, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to maligcy in breast cancer, suggesting that ZBTB8 may act as a transcriptional repressor or be involved in tumorigenesis.Spécification
ZBTB8A | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BR9 | |
BOZ F1, BOZF1, ZBTB8, ZBTB8A | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
653121 |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
ZBTB8A | |
Liquid | |
ZBTB8A | |
ZBTB8A Fusion Protein Ag21588 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |