missing translation for 'onlineSavingsMsg'
Learn More

ZDHHC1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26545-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16804975

  • 112.71€ / 20µL

Expédition estimée
26-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

26545-1-AP recognizes 48-55 kDa bands.

ZDHHC1 (Zinc Finger DHHC-Type Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include protein-cysteine S-palmitoyltransferase activity and palmitoyltransferase activity.
Spécification

Spécification

ZDHHC1
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WTX9
C16orf1, DHHC 1, HSU90653, ZDHHC1, Zinc finger protein 377, ZNF377
Rabbit
IgG
20 μL
-20°C
Primary
29800
Western Blot
Unconjugated
ZDHHC1
Liquid
ZDHHC1
ZDHHC1 Fusion Protein Ag24028
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents