missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFPL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27157-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ZFPL1 is expressed strongly in the exocrine pancreas as a 1.4-kb polyadenylated RNA encoding a putative protein of 310 amino acids.Spécification
ZFPL1 | |
Polyclonal | |
Unconjugated | |
ZFPL1 | |
D11S750, MCG4, ZFPL1, zinc finger protein like 1, Zinc finger protein MCG4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7542 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95159 | |
ZFPL1 | |
ZFPL1 Fusion Protein Ag24097 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |