missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF284 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24681-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ZNF284 may be involved in transcriptional regulation.Spécification
ZNF284 | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2VY69 | |
zinc finger protein 284, ZNF284, ZNF284L | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
342909 |
Western Blot | |
Unconjugated | |
ZNF284 | |
Liquid | |
ZNF284 | |
ZNF284 Fusion Protein Ag20336 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |