missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF48 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25463-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ZNF48, also named as zinc finger protein 48, is 618 amino acid protein, which contains twelves C2H2-type zinc fingers and belongs to the Krueppel C2H2-type zinc-finger protein family. ZNF48 localizes in the nucleus and may be involves in transcription regulation.Spécification
ZNF48 | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96MX3 | |
zinc finger protein 48, Zinc finger protein 553, ZNF48, ZNF553 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
197407 |
Western Blot | |
Unconjugated | |
ZNF48 | |
Liquid | |
ZNF48 | |
ZNF48 Fusion Protein Ag22208 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |