Media Bottles
Links
Save Now - Web Exclusive Deals
Take advantage of our web exclusive prices and place your order today.
DWK Life Sciences DURAN™ Original Laboratory Bottle
Corning™ PET Storage Bottles
Must Have
Complete your media bottles order with these supplies.
Complete Your Order
  • Great deals
    DWK Life Sciences DURAN™ YOUTILITY™ Bottle Tag, GL 45, Silicone Rubber, Assorted
    Bottle tags for DWK Life Sciences DURAN™ YOUTILITY™ GL 45 Bottles, Assorted Colors.
    Learn More
  • Great deals
    Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier
    Protect your bottles with this practical carrier.
    Learn More
  • Great deals
    DWK Life Sciences DURAN™ Original Laboratory GL Bottle Pouring Rings
    Cap plug sealing system to ensure a liquid tight seal, available in four colors.
    Learn More
Guides to Help You Choose
Focus on Glassware & Plasticware
Focus on Glassware & Plasticware
Your essential guide to everyday labware. Clear your doubts about different material types and learn how to care for your items. The guide also includes a chemical compatibility chart and a FAQ section.
Download
Thermo Scientific™ Nalgene™ Bottle and Carboy Technical Guide
Thermo Scientific™ Nalgene™ Bottle and Carboy Technical Guide
Get familiar with Nalgene™ products, certifications, and care instructions. The guide also includes a section about plastic resins and their compatibility with chemicals.
Download
DURAN™ Evolutionary Bottle System
DURAN™ Evolutionary Bottle System
An extensive introduction to DURAN™ bottles, caps, and accessories. Understand the main features of different bottle types and their application.
Discover Now
Shop Your Brands
Frequently Asked Questions
What is the best material for media bottles?
Plastic media bottles are lightweight and impact resistant. They are recommended for use when unbreakable containers are needed or for work at low temperatures. Glass bottles can withstand higher temperatures and they are also available in darker glass suitable for working with light-sensitive materials.
Can plastic bottles be autoclaved?
Generally, polypropylene, PTFE, PFA, and PMP materials can be autoclaved, but temperatures higher than 80°C can affect the accuracy of volumetric vessels. Always refer back to the product technical guide and remember to loosen any caps before autoclaving.
What kind of detergents can I use on my plastic bottles?
Ideally, a low or non-alkaline detergent should be used as some plastics such as polystyrene and polycarbonate are susceptible to alkalis. Avoid abrasive materials and wash below 60°C if using an electric washer. For further information check the product guide.
Browse Other Product Categories