CAS RN 313474-59-8
CAS RN 313474-59-8
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
16alpha,17-[(1RS)-Butylidenebis(oxy)]-9alpha-bromo-11beta,21-dihydroxypregna-1,4-diene-3,20-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
9a-Bromobudesonide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
9Alpha-Bromobudesonide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.