CAS RN 190436-05-6
CAS RN 190436-05-6
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress β-Amyloid (1-11)
MedChemExpress β-Amyloid (1-11) is a fragment of Amyloid-β peptide, maybe used in the research of neurological disease.
2
MedChemExpress β-Amyloid (1-11)
MedChemExpress β-Amyloid (1-11) is a fragment of Amyloid-β peptide, maybe used in the research of neurological disease.
3
MedChemExpress β-Amyloid (1-11)
MedChemExpress β-Amyloid (1-11) is a fragment of Amyloid-β peptide, maybe used in the research of neurological disease.