Learn More
1,1,1-Trifluoro-2-butanone, TRC
Product Code. 30455181
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455181 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455181 Supplier Toronto Research Chemicals Supplier No. TRCT3818805G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1,1-Trifluoro-2-butanone
CAS 381-88-4
Molecular Formula C4H5F3O
Quantity 5 g
Synonym Ethyl Trifluoromethyl Ketone, NSC 42602
InChI Formula InChI=1S/C4H5F3O/c1-2-3(8)4(5,6)7/h2H2,1H3
SMILES CCC(=O)C(F)(F)F
IUPAC Name 1,1,1-trifluorobutan-2-one
Molecular Weight (g/mol) 126.08
Formula Weight 126.0292