Learn More
[1,1'-Biphenyl]-3-ylmethanol, 98%, BLD Pharm™
Spécification
Spécification
| CAS | 69605-90-9 |
| Molecular Formula | C13H12O |
| MDL Number | MFCD08703631 |
| Quantity | 1 g |
| SMILES | OCC1=CC(C2=CC=CC=C2)=CC=C1 |
| Molecular Weight (g/mol) | 184.23 |
| Percent Purity | 98% |
| Chemical Name or Material | [1,1'-Biphenyl]-3-ylmethanol |