Learn More
1-(1-Hydroxynaphthalen-2-yl)ethan-1-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 711-79-5 |
| Molecular Formula | C12H10O2 |
| MDL Number | MFCD00003963 |
| Quantity | 100 g |
| SMILES | CC(C1=CC=C2C=CC=CC2=C1O)=O |
| Molecular Weight (g/mol) | 186.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(1-Hydroxynaphthalen-2-yl)ethan-1-one |