Learn More
1,2,3,4,5,6,7,8-Octahydroacridine, TRC
Product Code. 30409656
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409656 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409656 Supplier Toronto Research Chemicals Supplier No. TRCO148513500MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,4,5,6,7,8-Octahydroacridine
CAS 1658-08-8
Molecular Formula C13H17N
Quantity 500 mg
InChI Formula InChI=1S/C13H17N/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h9H,1-8H2
SMILES C1CCc2nc3CCCCc3cc2C1
IUPAC Name 1,2,3,4,5,6,7,8-octahydroacridine
Molecular Weight (g/mol) 187.28
Formula Weight 187.1361