Learn More
1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one, TRC
Product Code. 30420277
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420277 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420277 Supplier Toronto Research Chemicals Supplier No. TRCT29370025MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one
CAS 196597-78-1
Molecular Formula C11 H10 O2
Quantity 25 mg
Synonym Ramelteon Impurity 5
InChI Formula InChI=1S/C11H10O2/c12-9-3-1-7-2-4-10-8(11(7)9)5-6-13-10/h2,4H,1,3,5-6H2
SMILES O=C1CCc2ccc3OCCc3c12
IUPAC Name 1,2,6,7-tetrahydrocyclopenta[e][1]benzofuran-8-one
Molecular Weight (g/mol) 174.2
Formula Weight 174.0681