Learn More
1,3-Benzoxazol-2-amine, Mikromol™
Product Code. 17900796
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17900796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17900796 Supplier Mikromol Supplier No. MM0332.170025
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,3-Benzoxazol-2-amine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Benzoxazol-2-amine
CAS 4570-41-6
Analyte or Component Names 1,3-Benzoxazol-2-amine
Shipping Condition Room Temperature
Molecular Formula C7 H6 N2 O
Certifications/Compliance ISO 17025
Synonym 2-Benzoxazolamine, 1,3-Benzoxazol-2-amine
InChI Formula InChI=1S/C7H6N2O/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9)
SMILES Nc1oc2ccccc2n1
IUPAC Name 1,3-benzoxazol-2-amine
Molecular Weight (g/mol) 134.14
Quantity 25 mg
Formula Weight 134.048 g/mol