Learn More
1,3-Bis(2-ethylhexyl)-5-methylhexahydropyrimidin-5-amine, 98%, BLD Pharm™
Product Code. 31089335
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31089335 250 mg
31080962 1 g
31088404 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31089335 Supplier BLD Pharm Supplier No. BD133334250mg
 Restricted Item
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 141-94-6
Molecular Formula C21H45N3
MDL Number MFCD00010428
Quantity 250 mg
SMILES NC1(CN(CN(C1)CC(CCCC)CC)CC(CCCC)CC)C
Molecular Weight (g/mol) 339.60
Percent Purity 98%
Chemical Name or Material 1,3-Bis(2-ethylhexyl)-5-methylhexahydropyrimidin-5-amine