Learn More
1-(3-Bromophenyl)thiourea, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(3-Bromophenyl)thiourea |
| CAS | 21327-14-0 |
| Molecular Formula | C7H7BrN2S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H7BrN2S/c8-5-2-1-3-6(4-5)10-7(9)11/h1-4H,(H3,9,10,11) |
| SMILES | NC(=S)Nc1cccc(Br)c1 |
| IUPAC Name | (3-bromophenyl)thiourea |
| Molecular Weight (g/mol) | 231.11 |
| Formula Weight | 229.9513 |