Learn More
1,3-Dimethyluracil, 99%, Thermo Scientific Chemicals
Catalog No. 11375475
Ansicht ändern
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Unit Size
11375475 1 g 1g
11385475 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375475 Supplier Thermo Scientific Alfa Aesar Supplier No. L19664.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 874-14-6
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00038065
InChI Key JSDBKAHWADVXFU-UHFFFAOYSA-N
Synonym 1,3-dimethyluracil, n1,n3-dimethyluracil, n,n'-dimethyluracil, 1,3-dimethylpyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 1,3-dimethyl, 2,4-dihydroxy-1,3-dimethylpyrimidine, uracil, 1,3-dimethyl, 1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione, 1,3-dimethyl-1h-pyrimidine-2,4-dione
PubChem CID 70122
ChEBI CHEBI:74763
IUPAC Name 1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C=CC(=O)N(C)C1=O
Specifications
Melting Point 119°C to 125°C
Quantity 1 g
Beilstein 124074
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material 1,3-Dimethyluracil