Learn More
1,3-Diphenylurea, 95%, BLD Pharm™
Product Code. 31032965
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032965 100 g -
31028173 500 g -
31016200 25 g 25g
31022140 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31032965 Supplier BLD Pharm Supplier No. BD119957100g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 102-07-8
Molecular Formula C13H12N2O
MDL Number MFCD00003017
Quantity 100 g
SMILES O=C(NC1=CC=CC=C1)NC2=CC=CC=C2
Molecular Weight (g/mol) 212.25
Percent Purity 95%
Chemical Name or Material 1,3-Diphenylurea