Learn More
1-(3-Methylpyrazin-2-yl)ethanone, 98%, BLD Pharm™
Product Code. 30948729
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948729 1 g 1g
30940012 10 g 10g
30943501 5 g 5g
30945173 100 g 100g
30939791 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30948729 Supplier BLD Pharm Supplier No. BD105621g
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 23787-80-6
Molecular Formula C7H8N2O
MDL Number MFCD00014612
Quantity 1 g
SMILES CC(=O)C1=C(C)N=CC=N1
Molecular Weight (g/mol) 136.15
Percent Purity 98%
Chemical Name or Material 1-(3-Methylpyrazin-2-yl)ethanone