Learn More
1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under argon in resealable AcroSeal™ bottles
Product Code. 16368207
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
Unit Size:
1 L
100mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16368207 100 mL 100mL
16378207 1 L 1 L
16388207 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16368207 Supplier Thermo Scientific Alfa Aesar Supplier No. 047305.AE
  • ONLINE EXCLUSIVE PRICE
    61.70€
    52.65€ / 100mL
    Économisez 9.05€
    14% de réduction
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
CAS: 123-91-1 | C{4}H{8}O{2}
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 11.8°C
CAS 123-91-1
Quantity 100 mL
Molecular Formula C{4}H{8}O{2}
Boiling Point 100°C to 102°C
Percent Purity 99.8%